Search
Search

Dhdds| Gene Symbol: Dhdds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026144.4 | NP_080420.2 | 3 - 4 | 325 | 62 | |
| GenBank mRNA | AK004993.1 | - | 3 - 4 | 387 | 62 | |
| AK034277.1 | - | 3 - 4 | 387 | 62 | ||
| AK036929.1 | - | 3 - 4 | 403 | 62 | ||
| AK049991.1 | - | 3 - 4 | 326 | 62 | ||
| AK135087.1 | - | 3 - 4 | 387 | 62 | ||
| AK149138.1 | - | 3 - 4 | 357 | 62 | ||
| AK171207.1 | - | 3 - 4 | 399 | 62 | ||
| BC004011.1 | - | 3 - 4 | 373 | 3493585 | 62 | |
Set Membership: |
|