Search
Search

Rims3| Gene Symbol: Rims3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182929.2 | NP_891559.1 | 1 - 2 | 220 | 75 | |
| GenBank mRNA | AK043152.1 | - | 3 - 4 | 719 | 75 | |
| AK047745.1 | - | 3 - 4 | 733 | 75 | ||
| AK122226.2 | - | 1 - 2 | 88 | 75 | ||
| AK136831.1 | - | 3 - 4 | 512 | 75 | ||
| AY326953.1 | - | 1 - 2 | 220 | 75 | ||
| BC115536.2 | - | 1 - 2 | 220 | 40101461 | 75 | |
Set Membership: |
|