Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Eif2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111277.1 | NP_001104747.1 | 6 - 7 | 732 | 74 | |
NM_175135.4 | NP_780344.2 | 6 - 7 | 732 | 74 | ||
GenBank mRNA | AK145856.1 | - | 6 - 7 | 709 | 74 | |
AK172173.1 | - | 5 - 6 | 614 | 74 | ||
BC115657.1 | - | 4 - 5 | 406 | 40103205 | 74 | |
BC115658.1 | - | 4 - 5 | 406 | 40103206 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |