Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01187545_g1
          See other ARHGAP4 GE Assays ›
          Gene Symbol:
          Arhgap4
          Gene Name:
          Rho GTPase activating protein 4
          Gene Aliases:
          A130009C12Rik, A530015A20Rik, Rgc1, c1, mKIAA0131
          Chromosome Location:
          Chr.X: 73894352 - 73911346 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01187545_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arhgap4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001162423.1 NP_001155895.1 19 - 20 2479 64
          NM_001162424.1 NP_001155896.1 18 - 19 2449 64
          NM_138630.2 NP_619536.2 19 - 20 2512 64
          GenBank mRNA AF443826.1 - 19 - 20 2443 64
          AK040687.1 - 12 - 13 1318 64
          AK129060.1 - 3021 64
          AK157294.1 - 19 - 20 2511 64
          AK169865.1 - 16 - 17 1947 64
          AY183124.1 - 19 - 20 2485 64
          AY183125.1 - 18 - 19 2422 64
          AY183126.1 - 18 - 19 2579 64
          AY183127.1 - 19 - 20 2485 64
          AY183128.1 - 15 - 16 2174 64
          AY183129.1 - 19 - 20 2485 64
          AY183130.1 - 15 - 16 1857 64
          AY183131.1 - 14 - 15 1951 64
          AY183132.1 - 13 - 14 2177 64
          AY183133.1 - 17 - 18 2518 64
          BC109001.1 - 19 - 20 2410 40048081 64
          BC109002.1 - 17 - 18 2469 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003430.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arhgap4

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of fibroblast migration
          negative regulation of cell migration
          negative regulation of axon extension
          positive regulation of GTPase activity
          GTPase activator activity
          Rac GTPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00015195.mm.1 NM_001162423.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015195.mm.1 NM_001162423.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015195.mm.1 NM_001162424.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015195.mm.1 NM_001162424.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015195.mm.1 NM_138630.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015195.mm.1 NM_138630.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015195.mm.1 TC0X00003430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00015196.mm.1 NM_001162423.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015196.mm.1 NM_001162423.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015196.mm.1 NM_001162424.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015196.mm.1 NM_001162424.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015196.mm.1 NM_138630.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015196.mm.1 NM_138630.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015196.mm.1 TC0X00003430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00015191.mm.1 NM_001162423.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015191.mm.1 NM_001162423.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015191.mm.1 NM_001162424.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015191.mm.1 NM_001162424.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015191.mm.1 NM_138630.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015191.mm.1 NM_138630.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015191.mm.1 TC0X00003430.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline