Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01187594_g1
          See other GATSL3 GE Assays ›
          Gene Symbol:
          Gatsl3
          Gene Name:
          GATS protein-like 3
          Gene Aliases:
          2410008K03Rik
          Chromosome Location:
          Chr.11: 4218162 - 4222409 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01187594_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gatsl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028022.1 NP_082298.1 8 - 9 985 82
          GenBank mRNA AK010455.1 - 8 - 9 968 82
          BC039988.1 - 8 - 9 985 1195616 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000039.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gatsl3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27556424 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27556422 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27556423 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27556420 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27556421 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27556419 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P517051 BC039988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P517051 NM_028022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P517051 AK010455.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P517051 BC039988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P517051 NM_028022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P517051 AK010455.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 381955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 601623 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P517051 BC039988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P517051 NM_028022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P517051 AK010455.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P517051 BC039988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P517051 NM_028022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P517051 AK010455.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556412 - -
          Whole Mouse Genome G4122A A_51_P517051 BC039988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517051 NM_028022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517051 AK010455.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517051 BC039988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517051 NM_028022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517051 AK010455.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09655960 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08976792 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09655961 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08976791 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27556423 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27556422 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27556419 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27556424 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27556420 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27556421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09655959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08976798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09294553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09649561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09603824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09705888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09738128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09603828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09603850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556449 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373911 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373903 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373906 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373908 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373910 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373905 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373907 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373904 - -
          Affymetrix HT MG-430 PM Array Plate 1452847_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10373902 - -
          Mouse Genome 430 2.0 Array 1452847_PM_at - -
          Mouse Genome 430 2.0 Array 1452847_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71962_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71962_at - -
          Affymetrix Mouse Clariom S Array PSR1100000510.mm.1 NM_028022.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000510.mm.1 NM_028022.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000510.mm.1 TC1100000039.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000509.mm.1 NM_028022.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000509.mm.1 NM_028022.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000509.mm.1 TC1100000039.mm.1 MTA TC
          Mouse Expression Set 430 1452847_at - -
          Mouse Expression Set 430 1452847_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452847_at - -
          Affymetrix HT Mouse Genome 430B Array 1452847_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 677072 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2759563 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2759563 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline