Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01187695_g1
          See other UQCR10 GE Assays ›
          Gene Symbol:
          Uqcr10
          Gene Name:
          ubiquinol-cytochrome c reductase, complex III subunit X
          Gene Aliases:
          1110020P15Rik, AA960494, Ucrc
          Chromosome Location:
          Chr.11: 4701968 - 4704362 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01187695_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Uqcr10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_197979.2 NP_932096.1 1 - 2 184 86
          GenBank mRNA AK002227.1 - 1 - 2 174 86
          AK003881.1 - 1 - 2 181 86
          AK012489.1 - 1 - 2 177 86
          AK168491.1 - 1 - 2 158 86
          AW985484.1 - 1 - 2 152 86
          BC024518.1 - 1 - 2 168 4984172 86
          BC087874.1 - 1 - 2 174 6545811 86
          BC100591.1 - 1 - 2 174 30119194 86
          CB599515.1 - 1 - 2 187 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Uqcr10

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial electron transport, ubiquinol to cytochrome c
          transport
          biological_process
          oxidation-reduction process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166414 AW985484.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166414 BC024518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166414 NM_197979.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166414 AK002227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166414 AK003881.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136765 BC065793.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136765 BC085508.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136765 BI202508.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136765 BM937611.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136765 NR_015545.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157705 AW985484.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157705 NM_197979.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115620 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136765 BC065793.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136765 BC085508.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136765 BI202508.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136765 BM937611.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136765 NR_015545.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166414 AW985484.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166414 BC024518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166414 NM_197979.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166414 AK002227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166414 AK003881.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157705 AW985484.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157705 NM_197979.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27559088 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115620 - -
          Whole Mouse Genome G4122A A_51_P387334 AK168491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P387334 AW985484.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P387334 BC024518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P387334 BC087874.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P387334 NM_197979.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140412 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115620 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09218412 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09218419 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09218410 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09218418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27559095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09166967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09466476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09602400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09058438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09218419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09166965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09581642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09218414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09166968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27559078 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465044 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383888 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383890 - -
          Affymetrix HT MG-430 PM Array Plate 1424364_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465043 - -
          Affymetrix Mouse Gene 1.1 ST Array 10383887 - -
          Mouse Genome 430 2.0 Array 1424364_a_at - -
          Mouse Genome 430 2.0 Array 1424364_PM_a_at - -
          Mouse Genome 430 2.0 Array 1434341_PM_x_at - -
          Mouse Genome 430 2.0 Array 1434341_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66152_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66152_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100020575.mm.1 NM_197979.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020575.mm.1 NM_197979.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020575.mm.1 TC1100002119.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100020578.mm.1 NM_197979.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020578.mm.1 NM_197979.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020578.mm.1 TC1100002119.mm.1 MTA TC
          Mouse Expression Set 430 1434341_x_at - -
          Mouse Expression Set 430 1424364_PM_a_at - -
          Mouse Expression Set 430 1434340_PM_at - -
          Mouse Expression Set 430 1424364_a_at - -
          Mouse Expression Set 430 1434341_PM_x_at - -
          Mouse Expression Set 430 1434340_at - -
          Murine Genome U74v2 Set 94078_PM_at - -
          Murine Genome U74v2 Set 94078_at - -
          Mouse Genome 430A 2.0 Array 1424364_a_at - -
          Mouse Genome 430A 2.0 Array 1434341_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1434340_PM_at - -
          Mouse Genome 430A 2.0 Array 1434340_at - -
          Mouse Genome 430A 2.0 Array 1434341_x_at - -
          Mouse Genome 430A 2.0 Array 1424364_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424364_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424364_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 883725 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3520452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003520452 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2636384 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2636384 - -
          mouse-6 v1.1 (Illumina) scl40580.5.1_111-S - -
          Illumina Sentrix mouse-6.v1 bead array 3520452 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40580.5.1_111-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3520452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2636384 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3142224 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2636384 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3142224 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40580.5.1_111-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline