Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01187700_g1
          See other GAS2L1 GE Assays ›
          Gene Symbol:
          Gas2l1
          Gene Name:
          growth arrest-specific 2 like 1
          Gene Aliases:
          4930500E24Rik, D0Jmb1, Gar22, TU-71.1
          Chromosome Location:
          Chr.11: 5054131 - 5065327 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01187700_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gas2l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030228.3 NP_084504.2 1 - 2 113 145
          NM_144560.3 NP_653146.1 1 - 2 113 145
          GenBank mRNA AK019661.1 - 1 - 2 88 145
          AK052229.1 - 1 - 2 114 145
          CJ170523.1 - 1 - 2 83 145

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK019661.1, NM_030228.3, NM_144560.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gas2l1

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule bundle formation
          negative regulation of microtubule depolymerization
          cell cycle arrest
          cellular response to starvation
          negative regulation of gene expression
          negative regulation of cell growth
          negative regulation of erythrocyte differentiation
          regulation of cell cycle
          cellular response to thyroid hormone stimulus
          microtubule binding
          cytoskeletal adaptor activity
          thyroid hormone receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100020663.mm.1 NM_030228.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020663.mm.1 NM_030228.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020663.mm.1 NM_144560.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020663.mm.1 NM_144560.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020663.mm.1 TC1100002124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100020662.mm.1 NM_030228.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020662.mm.1 NM_030228.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020662.mm.1 NM_144560.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020662.mm.1 NM_144560.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020662.mm.1 TC1100002124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100020664.mm.1 NM_001190406.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020664.mm.1 NM_001190406.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020664.mm.1 NM_001190408.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020664.mm.1 NM_001190408.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020664.mm.1 NM_030228.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020664.mm.1 NM_030228.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020664.mm.1 NM_144560.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020664.mm.1 NM_144560.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020664.mm.1 TC1100002124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100020668.mm.1 NM_001190406.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020668.mm.1 NM_001190406.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020668.mm.1 NM_001190408.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020668.mm.1 NM_001190408.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020668.mm.1 NM_030228.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020668.mm.1 NM_030228.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020668.mm.1 NM_144560.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020668.mm.1 NM_144560.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020668.mm.1 TC1100002124.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline