Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01187857_m1
          See other EGFR GE Assays ›
          Gene Symbol:
          Egfr
          Gene Name:
          epidermal growth factor receptor
          Gene Aliases:
          9030024J15Rik, AI552599, Erbb, Errb1, Errp, Wa5, wa-2, wa2
          Chromosome Location:
          Chr.11: 16752203 - 16913907 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01187857_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Egfr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007912.4 NP_031938.1 13 - 14 1911 72
          NM_207655.2 NP_997538.1 13 - 14 1911 72
          GenBank mRNA AF124513.1 - 13 - 14 1845 72
          AF275367.1 - 13 - 14 1854 72
          AF277898.1 - 1 - 2 110 72
          AK004883.1 - 13 - 14 1912 72
          AK004911.1 - 13 - 14 1886 72
          AK004944.1 - 13 - 14 1905 72
          AK134262.1 - 13 - 14 1884 72
          BC023729.1 - 13 - 14 1882 5351807 72
          U03425.1 - 13 - 14 1681 72
          X78987.1 - 13 - 14 1690 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000161.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Egfr

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          cell morphogenesis
          embryonic placenta development
          positive regulation of protein phosphorylation
          hair follicle development
          translation
          protein phosphorylation
          signal transduction
          cell surface receptor signaling pathway
          transmembrane receptor protein tyrosine kinase signaling pathway
          epidermal growth factor receptor signaling pathway
          multicellular organism development
          salivary gland morphogenesis
          learning or memory
          cell proliferation
          positive regulation of cell proliferation
          epidermis development
          magnesium ion homeostasis
          phosphorylation
          single organismal cell-cell adhesion
          peptidyl-tyrosine phosphorylation
          cerebral cortex cell migration
          positive regulation of cell migration
          positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
          positive regulation of superoxide anion generation
          positive regulation of catenin import into nucleus
          intracellular signal transduction
          regulation of cell proliferation
          negative regulation of protein catabolic process
          positive regulation of phosphorylation
          ovulation cycle
          negative regulation of apoptotic process
          positive regulation of MAP kinase activity
          positive regulation of nitric oxide biosynthetic process
          positive regulation of DNA repair
          positive regulation of DNA replication
          positive regulation of vasoconstriction
          positive regulation of vasodilation
          negative regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          astrocyte activation
          positive regulation of fibroblast proliferation
          digestive tract morphogenesis
          positive regulation of smooth muscle cell proliferation
          neuron projection morphogenesis
          positive regulation of epithelial cell proliferation
          positive regulation of inflammatory response
          regulation of peptidyl-tyrosine phosphorylation
          regulation of nitric-oxide synthase activity
          response to calcium ion
          positive regulation of protein kinase B signaling
          positive regulation of synaptic transmission, glutamatergic
          morphogenesis of an epithelial fold
          response to UV-A
          positive regulation of ERK1 and ERK2 cascade
          cellular response to amino acid stimulus
          cellular response to epidermal growth factor stimulus
          cellular response to estradiol stimulus
          nucleotide binding
          glycoprotein binding
          chromatin binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          signal transducer activity
          transmembrane signaling receptor activity
          epidermal growth factor-activated receptor activity
          receptor binding
          integrin binding
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          enzyme binding
          protein kinase binding
          protein phosphatase binding
          nitric-oxide synthase regulator activity
          ubiquitin protein ligase binding
          identical protein binding
          protein heterodimerization activity
          epidermal growth factor binding
          actin filament binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100001451.mm.1 NM_007912.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001451.mm.1 NM_007912.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001451.mm.1 NM_207655.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001451.mm.1 NM_207655.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001451.mm.1 TC1100000161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100001450.mm.1 NM_007912.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001450.mm.1 NM_007912.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001450.mm.1 NM_207655.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001450.mm.1 NM_207655.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001450.mm.1 TC1100000161.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline