Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tcf7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079822.2 | NP_001073290.1 | 2 - 3 | 397 | 89 | |
NM_009332.3 | NP_033358.2 | 2 - 3 | 397 | 89 | ||
GenBank mRNA | AJ223233.1 | - | 2 - 3 | 220 | 89 | |
BC128305.1 | - | 2 - 3 | 304 | 40109611 | 89 | |
BC128306.1 | - | 2 - 3 | 304 | 40109613 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |