Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm01189441_m1
See other ALMS1 GE Assays ›
Gene Symbol:
Alms1
Gene Name:
Alstrom syndrome 1
Gene Aliases:
bbb
Chromosome Location:
Chr.6: 85587499 - 85726611 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm01189441_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Alms1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_145223.2
NP_660258.2
6 - 7
1186
77
GenBank mRNA
AF425257.1
-
6 - 7
1187
77
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC0600000964.mm.1
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mus musculus
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Alms1
Gene Ontology Categories:
Function(s)
Process(es)
cellular glucose homeostasis
establishment of planar polarity
lipid metabolic process
spermatid development
sensory perception of sound
endosomal transport
calcium-mediated signaling
ovulation
negative regulation of multicellular organism growth
cilium assembly
glucose homeostasis
cholesterol homeostasis
negative regulation of apoptotic process
fat cell differentiation
regulation of fat cell differentiation
retinal rod cell development
epithelial cell proliferation
regulation of stress fiber assembly
inner ear receptor stereocilium organization
alpha-actinin binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR0600007457.mm.1
NM_145223.2.1
Refseq
Affymetrix Mouse Clariom S Array
PSR0600007457.mm.1
NM_145223.2
Refseq
Affymetrix Mouse Clariom S Array
PSR0600007457.mm.1
TC0600000964.mm.1
MTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit