Search
Search

Slc35f1| Gene Symbol: Slc35f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178675.4 | NP_848790.2 | 4 - 5 | 1124 | 73 | |
| GenBank mRNA | AK038601.1 | - | 4 - 5 | 1119 | 73 | |
| AK048957.1 | - | 4 - 5 | 1084 | 73 | ||
| AK053426.1 | - | 4 - 5 | 1093 | 73 | ||
| AK053545.1 | - | 4 - 5 | 1063 | 73 | ||
| AK083768.1 | - | 4 - 5 | 1063 | 73 | ||
| AK139331.1 | - | 4 - 5 | 1094 | 73 | ||
| AK163806.1 | - | 4 - 5 | 924 | 73 | ||
| AK165021.1 | - | 4 - 5 | 1078 | 73 | ||
| AK166268.1 | - | 4 - 5 | 1052 | 73 | ||
| BC059075.1 | - | 4 - 5 | 1020 | 6826829 | 73 | |
Set Membership: |
|