Search
Search

Slc6a6| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009320.4 | NP_033346.2 | 8 - 9 | 1261 | 68 | |
| GenBank mRNA | AK143510.1 | - | 8 - 9 | 1225 | 68 | |
| BC015245.1 | - | 8 - 9 | 1198 | 4242193 | 68 | |
| L03292.2 | - | 8 - 9 | 1233 | 68 | ||
Set Membership: |
|