Search
Search

Nipal3| Gene Symbol: Nipal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028995.3 | NP_083271.1 | 4 - 5 | 668 | 97 | |
| GenBank mRNA | AK049696.1 | - | 4 - 5 | 703 | 97 | |
| AK053937.1 | - | 671 | 97 | |||
| AK082376.1 | - | 670 | 97 | |||
| AK087811.1 | - | 4 - 5 | 671 | 97 | ||
| AK138843.1 | - | 4 - 5 | 669 | 97 | ||
| BC057168.1 | - | 4 - 5 | 608 | 4973200 | 97 | |
Set Membership: |
|