Search
Search
| Gene Symbol: Fgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010208.4 | NP_034338.3 | 5 - 6 | 642 | 76 | |
| GenBank mRNA | AK036438.1 | - | 5 - 6 | 564 | 76 | |
| AK036476.1 | - | 5 - 6 | 814 | 76 | ||
| AK038141.1 | - | 5 - 6 | 548 | 76 | ||
| AK089812.1 | - | 5 - 6 | 612 | 76 | ||
| AK155902.1 | - | 5 - 6 | 618 | 76 | ||
| AK157640.1 | - | 596 | 76 | |||
| AK171824.1 | - | 5 - 6 | 596 | 76 | ||
| BC057863.1 | - | 5 - 6 | 511 | 5317449 | 76 | |
| X52191.1 | - | 5 - 6 | 549 | 76 | ||
Set Membership: |
|