Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01191612_g1
          See other SGK1 GE Assays ›
          Gene Symbol:
          Sgk1
          Gene Name:
          serum/glucocorticoid regulated kinase 1
          Gene Aliases:
          Sgk
          Chromosome Location:
          Chr.10: 21882184 - 21999903 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01191612_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sgk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001161845.2 NP_001155317.2 9 - 10 1391 73
          NM_001161847.2 NP_001155319.1 7 - 8 892 73
          NM_001161848.2 NP_001155320.1 7 - 8 685 73
          NM_001161849.2 NP_001155321.1 7 - 8 766 73
          NM_001161850.2 NP_001155322.1 7 - 8 873 73
          NM_011361.3 NP_035491.1 7 - 8 741 73
          GenBank mRNA AF139638.1 - 7 - 8 725 73
          AF205855.1 - 7 - 8 722 73
          AK086892.1 - 7 - 8 725 73
          AK132234.1 - 7 - 8 813 73
          AK146037.1 - 7 - 8 725 73
          AK146062.1 - 7 - 8 722 73
          AK159131.1 - 7 - 8 725 73
          AK167364.1 - 7 - 8 722 73
          BC005720.1 - 7 - 8 702 3594892 73
          BC070401.1 - 9 - 10 1391 5695396 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000171.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sgk1

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          cellular sodium ion homeostasis
          apoptotic process
          cellular response to DNA damage stimulus
          microtubule depolymerization
          long-term memory
          visual learning
          positive regulation of sodium ion transport
          phosphorylation
          peptidyl-serine phosphorylation
          positive regulation of cell growth
          negative regulation of microtubule polymerization
          cellular response to insulin stimulus
          regulation of protein localization
          negative regulation of apoptotic process
          glucocorticoid mediated signaling pathway
          neuron projection morphogenesis
          positive regulation of dendrite morphogenesis
          regulation of cell cycle
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          3-phosphoinositide-dependent protein kinase binding
          cofactor binding
          tau protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 NM_001161845.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 NM_001161845.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 NM_001161847.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 NM_001161847.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 NM_001161848.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 NM_001161848.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 NM_001161849.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 NM_001161849.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 NM_001161850.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 NM_001161850.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 NM_011361.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 NM_011361.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001258.mm.1 TC1000000171.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline