Search
Search
| Gene Symbol: Mecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025297.2 | NP_079573.2 | 5 - 6 | 692 | 70 | |
| GenBank mRNA | BC003864.1 | - | 5 - 6 | 692 | 3488760 | 70 |
| BQ746166.1 | - | 2 - 3 | 133 | 70 | ||
Set Membership: |
|