Search
Search

Ppil4| Gene Symbol: Ppil4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026141.3 | NP_080417.2 | 6 - 7 | 616 | 97 | |
| GenBank mRNA | AK011443.1 | - | 6 - 7 | 588 | 97 | |
| AK012371.1 | - | 6 - 7 | 593 | 97 | ||
| AK014406.2 | - | 589 | 97 | |||
| AK014453.1 | - | 5 - 6 | 1141 | 97 | ||
| AK017370.1 | - | 6 - 7 | 616 | 97 | ||
| AK167168.1 | - | 6 - 7 | 581 | 97 | ||
| AK167258.1 | - | 6 - 7 | 588 | 97 | ||
| BC079912.1 | - | 6 - 7 | 578 | 30661438 | 97 | |
Set Membership: |
|