Search
Search

Lats1| Gene Symbol: Lats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010690.1 | NP_034820.1 | 5 - 6 | 2957 | 85 | |
| GenBank mRNA | AF104414.1 | - | 2 - 3 | 2082 | 85 | |
| BC158092.1 | - | 4 - 5 | 2803 | 9088177 | 85 | |
| BC158123.1 | - | 5 - 6 | 2952 | 9088196 | 85 | |
Set Membership: |
|