| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400027654.mm.1 |
NM_013848.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027654.mm.1 |
NM_013848.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027654.mm.1 |
TC0400003418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027655.mm.1 |
NM_013848.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027655.mm.1 |
NM_013848.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027655.mm.1 |
TC0400003418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027657.mm.1 |
NM_013848.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027657.mm.1 |
NM_013848.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027657.mm.1 |
TC0400003418.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027656.mm.1 |
NM_013848.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027656.mm.1 |
NM_013848.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027656.mm.1 |
TC0400003418.mm.1 |
MTA TC |