| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100022989.mm.1 |
NM_010117.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022989.mm.1 |
NM_010117.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022989.mm.1 |
TC1100002471.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100022992.mm.1 |
NM_010117.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022992.mm.1 |
NM_010117.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022992.mm.1 |
TC1100002471.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100022985.mm.1 |
NM_010117.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022985.mm.1 |
NM_010117.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022985.mm.1 |
TC1100002471.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100022990.mm.1 |
NM_010117.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022990.mm.1 |
NM_010117.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022990.mm.1 |
TC1100002471.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100022986.mm.1 |
NM_010117.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022986.mm.1 |
NM_010117.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022986.mm.1 |
TC1100002471.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100022987.mm.1 |
NM_010117.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022987.mm.1 |
NM_010117.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022987.mm.1 |
TC1100002471.mm.1 |
MTA TC |