Search
Search

Rhbdf1| Gene Symbol: Rhbdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291818.1 | NP_001278747.1 | 9 - 10 | 1356 | 77 | |
| NM_010117.2 | NP_034247.2 | 9 - 10 | 1475 | 77 | ||
| GenBank mRNA | AK144437.1 | - | 9 - 10 | 1344 | 77 | |
| AK220566.1 | - | 6 - 7 | 2891 | 77 | ||
| BC023469.1 | - | 4 - 5 | 623 | 77 | ||
| BC027346.1 | - | 9 - 10 | 1420 | 4011421 | 77 | |
Set Membership: |
|