Search
Search

Lmtk3| Gene Symbol: Lmtk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005511.3 | NP_001005511.2 | 4 - 5 | 428 | 102 | |
| NM_001290990.1 | NP_001277919.1 | 3 - 4 | 583 | 102 | ||
| NR_110987.1 | 4 - 5 | 428 | 102 | |||
| GenBank mRNA | AB288873.1 | - | 4 - 5 | 422 | 102 | |
| BC057032.1 | - | 3 - 4 | 316 | 102 | ||
| BC059845.1 | - | 4 - 5 | 637 | 102 | ||
| BC083185.1 | - | 3 - 4 | 583 | 6855048 | 102 | |
| BC094377.1 | - | 4 - 5 | 428 | 30536822 | 102 | |
Set Membership: |
|