Search
Search

Cuedc2| Gene Symbol: Cuedc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164290.1 | NP_001157762.1 | 7 - 8 | 800 | 127 | |
| NM_001164291.1 | NP_001157763.1 | 6 - 7 | 692 | 127 | ||
| NM_001164292.1 | NP_001157764.1 | 6 - 7 | 689 | 127 | ||
| NM_001164293.1 | NP_001157765.1 | 6 - 7 | 659 | 127 | ||
| NM_001164294.1 | NP_001157766.1 | 6 - 7 | 638 | 127 | ||
| NM_001164295.1 | NP_001157767.1 | 6 - 7 | 635 | 127 | ||
| NM_024192.2 | NP_077154.2 | 7 - 8 | 791 | 127 | ||
| GenBank mRNA | AK008842.1 | - | 6 - 7 | 655 | 127 | |
| AK013884.1 | - | 6 - 7 | 650 | 127 | ||
| AK183087.1 | - | 1 - 2 | 172 | 127 | ||
| AK186847.1 | - | 1 - 2 | 174 | 126 | ||
Set Membership: |
|