Search
Search
| Gene Symbol: A1bg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081067.1 | NP_001074536.1 | 1 - 2 | 48 | 78 | |
| GenBank mRNA | BC080745.1 | - | 383 | 78 | ||
| DQ520794.1 | - | 1 - 2 | 44 | 78 | ||
Set Membership: |
|