| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10428971 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10428968 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10428966 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10428970 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10428969 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10428963 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10428962 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10428967 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10428965 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10428961 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10428964 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10428972 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10428960 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1500012157.mm.1 |
NM_183194.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012157.mm.1 |
NM_183194.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012157.mm.1 |
TC1500001622.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012151.mm.1 |
NM_183194.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012151.mm.1 |
NM_183194.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012151.mm.1 |
TC1500001622.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012155.mm.1 |
NM_183194.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012155.mm.1 |
NM_183194.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012155.mm.1 |
TC1500001622.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012156.mm.1 |
NM_183194.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012156.mm.1 |
NM_183194.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012156.mm.1 |
TC1500001622.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012152.mm.1 |
NM_183194.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012152.mm.1 |
NM_183194.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012152.mm.1 |
TC1500001622.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012154.mm.1 |
NM_183194.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012154.mm.1 |
NM_183194.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012154.mm.1 |
TC1500001622.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012153.mm.1 |
NM_183194.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012153.mm.1 |
NM_183194.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012153.mm.1 |
TC1500001622.mm.1 |
MTA TC |