Search
Search

Micall2| Gene Symbol: Micall2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174850.3 | NP_777275.2 | 14 - 15 | 2964 | 161 | |
| GenBank mRNA | AB182579.1 | - | 14 - 15 | 2815 | 161 | |
| AK044547.1 | - | 14 - 15 | 2816 | 161 | ||
| AK151086.1 | - | 14 - 15 | 2963 | 161 | ||
| AK164043.1 | - | 14 - 15 | 2935 | 161 | ||
| AK165558.1 | - | 14 - 15 | 2814 | 161 | ||
| AK220308.1 | - | 14 - 15 | 2741 | 161 | ||
| BC043300.1 | - | 14 - 15 | 2778 | 5025620 | 161 | |
Set Membership: |
|