Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01195270_g1
          See other CSF2RB2 GE Assays ›
          Gene Symbol:
          Csf2rb2
          Gene Name:
          colony stimulating factor 2 receptor, beta 2, low-affinity (granulocyte-macrophage)
          Gene Aliases:
          AIC2A, BetaIl3, Bil3, Csfgmrb, Il3r, Il3rb, Il3rb2
          Chromosome Location:
          Chr.15: 78282507 - 78305721 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01195270_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csf2rb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001287389.1 NP_001274318.1 9 - 10 1370 68
          NM_007781.3 NP_031807.1 11 - 12 1697 68
          GenBank mRNA AK154650.1 - 11 - 12 1620 68
          AK155216.1 - 11 - 12 1623 68
          AK155511.1 - 11 - 12 1599 68
          AK155626.1 - 9 - 10 1272 68
          AK155675.1 - 11 - 12 1599 68
          AK170028.1 - 11 - 12 1599 68
          M29855.1 - 11 - 12 1579 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001828.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csf2rb2

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine-mediated signaling pathway
          cytokine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2034663 M29855.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2034663 AK155626.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2034663 NM_007781.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146753 M29855.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146753 NM_007781.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146753 M29855.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P146753 NM_007781.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 534385 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 433409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58843 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 425910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149742 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 486036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733631 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142159 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111969 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105528 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154362 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2034663 M29855.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2034663 AK155626.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2034663 NM_007781.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146753 M29855.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146753 NM_007781.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146753 M29855.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P146753 NM_007781.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P146753 M29855.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P146753 NM_007781.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P146753 M29855.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P146753 NM_007781.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146753 M29855.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146753 NM_007781.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146753 M29855.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146753 NM_007781.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116410 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11841864 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11841860 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12423641 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11841861 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11841862 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11841851 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10430307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430303 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430305 - -
          Affymetrix HT MG-430 PM Array Plate 1449360_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10430302 - -
          Mouse Genome 430 2.0 Array 1449360_PM_at - -
          Mouse Genome 430 2.0 Array 1449360_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12984_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12984_at - -
          Affymetrix Mouse Clariom S Array PSR1500014282.mm.1 NM_001287389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014282.mm.1 NM_007781.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014282.mm.1 TC1500001828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014276.mm.1 NM_001287389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014276.mm.1 NM_007781.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014276.mm.1 TC1500001828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014278.mm.1 NM_001287389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014278.mm.1 NM_007781.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014278.mm.1 NR_106137.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014278.mm.1 NR_106138.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014278.mm.1 TC1500001828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014279.mm.1 NM_001287389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014279.mm.1 NM_007781.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014279.mm.1 TC1500001828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014280.mm.1 NM_001287389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014280.mm.1 NM_007781.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014280.mm.1 TC1500001828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014281.mm.1 NM_001287389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014281.mm.1 NM_007781.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014281.mm.1 TC1500001828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014277.mm.1 NM_001287389.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014277.mm.1 NM_007781.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014277.mm.1 TC1500001828.mm.1 MTA TC
          Mouse Expression Set 430 1450200_PM_s_at - -
          Mouse Expression Set 430 1450200_s_at - -
          Mouse Expression Set 430 1449360_at - -
          Mouse Expression Set 430 1449360_PM_at - -
          Murine Genome U74v2 Set 103210_at - -
          Murine Genome U74v2 Set 94748_PM_g_at - -
          Murine Genome U74v2 Set 103210_PM_at - -
          Murine Genome U74v2 Set 161544_PM_r_at - -
          Murine Genome U74v2 Set 161544_r_at - -
          Murine Genome U74v2 Set 94748_g_at - -
          Mouse Genome 430A 2.0 Array 1449360_at - -
          Mouse Genome 430A 2.0 Array 1450200_s_at - -
          Mouse Genome 430A 2.0 Array 1450200_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1449360_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449360_at - -
          Affymetrix HT Mouse Genome 430A Array 1449360_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 486066 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3120546 - -
          Illumina mouse-6 v1.0 expression beadchip 3120546 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120546 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2750047 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2750047 - -
          mouse-6 v1.1 (Illumina) scl46994.14_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 3120546 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46994.14_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120546 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2750047 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2750047 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline