Search
Search

Auts2| Gene Symbol: Auts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177047.3 | NP_796021.2 | 12 - 13 | 2424 | 79 | |
| GenBank mRNA | AK147478.1 | - | 3423 | 79 | ||
| AK164158.1 | - | 8 - 9 | 1864 | 79 | ||
| BC021509.1 | - | 5 - 6 | 499 | 79 | ||
| BC066072.1 | - | 7 - 8 | 2197 | 79 | ||
| BC096021.1 | - | 12 - 13 | 2423 | 79 | ||
Set Membership: |
|