Search
Search

Radil| Gene Symbol: Radil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289588.1 | NP_001276517.1 | 1 - 2 | 117 | 85 | |
| NM_178702.4 | NP_848817.2 | 1 - 2 | 117 | 85 | ||
| GenBank mRNA | AK052964.1 | - | 1 - 2 | 28 | 85 | |
| AK154169.1 | - | 1 - 2 | 56 | 85 | ||
| AK162917.1 | - | 1 - 2 | 97 | 85 | ||
| AK163852.1 | - | 1 - 2 | 53 | 85 | ||
| AK173277.1 | - | 1 - 2 | 81 | 85 | ||
| BC056483.1 | - | 1 - 2 | 69 | 5696312 | 85 | |
| BY201066.1 | - | 1 - 2 | 119 | 85 | ||
Set Membership: |
|