Search
Search
| Gene Symbol: Tbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011535.3 | NP_035665.2 | 1 - 2 | 1171 | 137 | |
| NM_198052.2 | NP_932169.1 | 2 - 3 | 1367 | 137 | ||
| GenBank mRNA | AK052453.1 | - | 1 - 2 | 1386 | 137 | |
| AK145436.1 | - | 1 - 2 | 1088 | 137 | ||
| AK157247.1 | - | 1 - 2 | 932 | 137 | ||
| AK172118.1 | - | 1 - 2 | 933 | 137 | ||
| BC096551.1 | - | 2 - 3 | 1367 | 30547736 | 137 | |
Set Membership: |
|