Search
Search
| Gene Symbol: Helq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081107.1 | NP_001074576.1 | 10 - 11 | 2195 | 103 | |
| GenBank mRNA | AF436846.1 | - | 8 - 9 | 1177 | 103 | |
| AK052427.1 | - | 10 - 11 | 2155 | 103 | ||
| BC082601.1 | - | 10 - 11 | 2159 | 30662103 | 103 | |
| BC109169.2 | - | 10 - 11 | 2063 | 103 | ||
| BC109170.2 | - | 10 - 11 | 2068 | 40048258 | 103 | |
Set Membership: |
|