Search
Search

Fbrsl1| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142642.1 | NP_001136114.1 | 16 - 17 | 2306 | 71 | |
| NM_028596.2 | NP_082872.2 | 9 - 10 | 1350 | 71 | ||
| GenBank mRNA | AK031967.1 | - | 9 - 10 | 1320 | 71 | |
| AK129388.1 | - | 5 - 6 | 2862 | 71 | ||
| BC059932.1 | - | 14 - 15 | 3092 | 71 | ||
| BC069994.1 | - | 4 - 5 | 307 | 71 | ||
| BC151196.1 | - | 11 - 12 | 1315 | 9088084 | 71 | |
| BU614458.1 | - | 6 - 7 | 542 | 71 | ||
Set Membership: |
|