Search
Search

Ddx55| Gene Symbol: Ddx55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190795.1 | NP_001177724.1 | 4 - 5 | 412 | 59 | |
| NM_026409.4 | NP_080685.2 | 4 - 5 | 412 | 59 | ||
| GenBank mRNA | AK012782.1 | - | 4 - 5 | 392 | 59 | |
| AK017554.1 | - | 4 - 5 | 354 | 59 | ||
| AK033741.1 | - | 4 - 5 | 389 | 59 | ||
| AK077525.1 | - | 4 - 5 | 393 | 59 | ||
| AK129402.1 | - | 4 - 5 | 390 | 59 | ||
| AK154422.1 | - | 4 - 5 | 413 | 59 | ||
| AK167149.1 | - | 3 - 4 | 2407 | 59 | ||
| BC043052.1 | - | 4 - 5 | 362 | 59 | ||
| BC117786.1 | - | 4 - 5 | 374 | 40092124 | 59 | |
| BC117787.1 | - | 4 - 5 | 374 | 40092125 | 59 | |
Set Membership: |
|