Search
Search

Asah2| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018830.1 | NP_061300.1 | 8 - 9 | 1054 | 121 | |
| GenBank mRNA | AB037111.1 | - | 9 - 10 | 1339 | 121 | |
| AB037181.1 | - | 8 - 9 | 1054 | 121 | ||
| AK034493.1 | - | 8 - 9 | 1285 | 121 | ||
| AK046540.1 | - | 8 - 9 | 1248 | 121 | ||
| AK047692.1 | - | 8 - 9 | 1242 | 121 | ||
| AK080951.1 | - | 8 - 9 | 1231 | 121 | ||
| AK166100.1 | - | 8 - 9 | 1302 | 121 | ||
| BC137898.1 | - | 8 - 9 | 1037 | 8860917 | 121 | |
| BC145016.1 | - | 8 - 9 | 1040 | 9053564 | 121 | |
Set Membership: |
|