Search
Search
| Gene Symbol: Sqle | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009270.3 | NP_033296.1 | 8 - 9 | 2094 | 106 | |
| GenBank mRNA | AK041198.1 | - | 1 - 2 | 635 | 106 | |
| AK138457.1 | - | 8 - 9 | 2107 | 106 | ||
| AK148419.1 | - | 8 - 9 | 2079 | 106 | ||
| AK161837.1 | - | 7 - 8 | 1984 | 106 | ||
| AK163502.1 | - | 8 - 9 | 2074 | 106 | ||
| AK180804.1 | - | 3 - 4 | 269 | 106 | ||
| AK189403.1 | - | 2 - 3 | 182 | 106 | ||
| AK208956.1 | - | 2 - 3 | 182 | 106 | ||
| AK209538.1 | - | 2 - 3 | 182 | 106 | ||
| BC042781.1 | - | 8 - 9 | 1351 | 4915911 | 106 | |
| BC056361.1 | - | 8 - 9 | 2094 | 6816800 | 106 | |
Set Membership: |
|