Search
Search

Rnft2| Gene Symbol: Rnft2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109902.1 | NP_001103372.1 | 7 - 8 | 1092 | 67 | |
| NM_172998.3 | NP_766586.2 | 8 - 9 | 1127 | 67 | ||
| GenBank mRNA | AK046850.1 | - | 8 - 9 | 1128 | 67 | |
| AK090256.1 | - | 7 - 8 | 1188 | 67 | ||
| AK148930.1 | - | 7 - 8 | 1069 | 67 | ||
| BC057302.1 | - | 7 - 8 | 1054 | 6827287 | 67 | |
Set Membership: |
|