Search
Search

Gnat2| Gene Symbol: Gnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008141.3 | NP_032167.1 | 6 - 7 | 673 | 71 | |
| GenBank mRNA | AK009388.1 | - | 5 - 6 | 557 | 71 | |
| AK028395.1 | - | 2 - 3 | 190 | 71 | ||
| BC016272.1 | - | 6 - 7 | 671 | 4506728 | 71 | |
| BC085102.1 | - | 4 - 5 | 1420 | 71 | ||
| BC131941.1 | - | 6 - 7 | 668 | 40130218 | 71 | |
| BC131943.1 | - | 6 - 7 | 668 | 40130220 | 71 | |
| L10666.1 | - | 5 - 6 | 595 | 71 | ||
Set Membership: |
|