Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01200536_g1
          See other PYCRL GE Assays ›
          Gene Symbol:
          Pycrl
          Gene Name:
          pyrroline-5-carboxylate reductase-like
          Gene Aliases:
          1110058B13Rik, 2700073G24Rik
          Chromosome Location:
          Chr.15: 75916463 - 75921560 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01200536_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pycrl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025412.2 NP_079688.2 5 - 6 736 59
          GenBank mRNA AK002912.1 - 5 - 6 667 59
          AK004291.1 - 5 - 6 740 59
          AK013214.1 - 5 - 6 661 59
          BC026536.1 - 5 - 6 657 4987954 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001753.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pycrl

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          proline biosynthetic process
          cellular amino acid biosynthetic process
          cytidine deamination
          oxidation-reduction process
          cytidine deaminase activity
          pyrroline-5-carboxylate reductase activity
          protein binding
          oxidoreductase activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30344973 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30344972 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30344971 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2153296 AK013214.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2153296 BC026536.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2153296 NM_025412.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2153296 AK002912.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126094 AK004291.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 781851 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 629726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 244 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177563 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126094 AK004291.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2153296 AK013214.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2153296 BC026536.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2153296 NM_025412.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2153296 AK002912.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30344966 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08420687 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177563 - -
          Whole Mouse Genome G4122A A_51_P482844 AK004291.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P179698 AK013214.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P179698 BC026536.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P179698 CX212991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P179698 NM_025412.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P179698 AK002912.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126094 AK004291.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177563 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12055632 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12297226 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11846229 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12122747 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12055634 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30344972 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30344973 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30344971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30345038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30344907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12377309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12377305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12055633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12377308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12284301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12297226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11846225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12055632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12284302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12206870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12055634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30344957 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10429671 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429672 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429668 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429669 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429667 - -
          Affymetrix HT MG-430 PM Array Plate 1416400_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10429666 - -
          Mouse Genome 430 2.0 Array 1416400_at - -
          Mouse Genome 430 2.0 Array 1416400_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66194_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66194_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500013255.mm.1 NM_025412.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013255.mm.1 NM_025412.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013255.mm.1 TC1500001753.mm.1 MTA TC
          Mouse Expression Set 430 1416400_at - -
          Mouse Expression Set 430 1416400_PM_at - -
          Murine Genome U74v2 Set 160197_at - -
          Murine Genome U74v2 Set 160197_PM_at - -
          Murine Genome U74v2 Set 166965_at - -
          Murine Genome U74v2 Set 166965_PM_at - -
          Mouse Genome 430A 2.0 Array 1416400_PM_at - -
          Mouse Genome 430A 2.0 Array 1416400_at - -
          Affymetrix HT Mouse Genome 430A Array 1416400_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416400_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 910155 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4780204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004780204 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775255 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775255 - -
          mouse-6 v1.1 (Illumina) scl066194.1_36-S - -
          Illumina Sentrix mouse-6.v1 bead array 4780204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066194.1_36-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4780204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228613 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228613 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066194.1_36-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline