Search
Search
| Gene Symbol: Fat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081286.2 | NP_001074755.2 | 15 - 16 | 10219 | 61 | |
| GenBank mRNA | AJ250768.1 | - | 15 - 16 | 10219 | 61 | |
| BC041794.1 | - | 1 - 2 | 72 | 61 | ||
| BC100554.1 | - | 2 - 3 | 338 | 61 | ||
Set Membership: |
|