Search
Search

Styxl1| Gene Symbol: Styxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289554.1 | NP_001276483.1 | 6 - 7 | 750 | 121 | |
| NM_001289555.1 | NP_001276484.1 | 6 - 7 | 747 | 121 | ||
| NM_001289556.1 | NP_001276485.1 | 5 - 6 | 591 | 121 | ||
| NM_029659.4 | NP_083935.3 | 6 - 7 | 757 | 121 | ||
| GenBank mRNA | AK005599.1 | - | 6 - 7 | 748 | 121 | |
| AK005858.1 | - | 5 - 6 | 498 | 121 | ||
| AK161310.1 | - | 6 - 7 | 758 | 121 | ||
| BC045148.1 | - | 6 - 7 | 687 | 6307739 | 121 | |
Set Membership: |
|