Search
Search

Nfkb2| Gene Symbol: Nfkb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177369.1 | NP_001170840.1 | 7 - 8 | 663 | 126 | |
| NM_001177370.1 | NP_001170841.1 | 6 - 7 | 579 | 126 | ||
| NM_019408.3 | NP_062281.1 | 6 - 7 | 814 | 126 | ||
| GenBank mRNA | AF155372.1 | - | 7 - 8 | 741 | 126 | |
| AF155373.1 | - | 7 - 8 | 778 | 126 | ||
| AK134081.1 | - | 7 - 8 | 1082 | 126 | ||
| AK137680.1 | - | 7 - 8 | 653 | 126 | ||
| AK144994.1 | - | 7 - 8 | 656 | 126 | ||
| AK145312.1 | - | 6 - 7 | 780 | 126 | ||
| AK148160.1 | - | 7 - 8 | 651 | 126 | ||
| AK150012.1 | - | 7 - 8 | 642 | 126 | ||
| AK153412.1 | - | 2 - 3 | 223 | 126 | ||
| BC027423.1 | - | 6 - 7 | 580 | 3672913 | 126 | |
| DQ020182.1 | - | 6 - 7 | 511 | 126 | ||
| DQ020183.1 | - | 5 - 6 | 632 | 126 | ||
| DQ020184.1 | - | 5 - 6 | 632 | 126 | ||
| DQ020185.1 | - | 6 - 7 | 511 | 126 | ||
Set Membership: |
|