Search
Search

Hnrnpd| Gene Symbol: Hnrnpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077265.2 | NP_001070733.1 | 5 - 6 | 1056 | 89 | |
| NM_001077266.2 | NP_001070734.1 | 4 - 5 | 999 | 89 | ||
| NM_001077267.2 | NP_001070735.1 | 4 - 5 | 999 | 89 | ||
| NM_007516.3 | NP_031542.2 | 5 - 6 | 1056 | 89 | ||
| GenBank mRNA | AK077409.1 | - | 5 - 6 | 1044 | 89 | |
| BC011172.1 | - | 5 - 6 | 731 | 89 | ||
| BC049098.1 | - | 5 - 6 | 1056 | 5717286 | 89 | |
| CA451481.1 | - | 3 - 4 | 337 | 89 | ||
| CF751545.1 | - | 2 - 3 | 207 | 89 | ||
Set Membership: |
|