Search
Search
| Gene Symbol: Dlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194940.2 | NP_001181869.1 | 11 - 12 | 3822 | 63 | |
| NM_001194941.1 | NP_001181870.1 | 7 - 8 | 2452 | 63 | ||
| NM_015802.3 | NP_056617.2 | 7 - 8 | 2370 | 63 | ||
| GenBank mRNA | AF178078.1 | - | 7 - 8 | 2306 | 63 | |
| AK147539.1 | - | 7 - 8 | 2452 | 63 | ||
| AK162413.1 | - | 7 - 8 | 2371 | 63 | ||
| AK166175.1 | - | 7 - 8 | 2330 | 63 | ||
| BC150802.1 | - | 7 - 8 | 2155 | 9087713 | 63 | |
| HM008381.1 | - | 10 - 11 | 3375 | 63 | ||
Set Membership: |
|