Search
Search
| Gene Symbol: Tchp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029992.2 | NP_084268.2 | 3 - 4 | 577 | 77 | |
| GenBank mRNA | AK020912.1 | - | 3 - 4 | 540 | 77 | |
| AK083027.1 | - | 3 - 4 | 493 | 77 | ||
| AK159947.1 | - | 3 - 4 | 538 | 77 | ||
| BC043048.1 | - | 3 - 4 | 482 | 5704036 | 77 | |
| BU512279.1 | - | 3 - 4 | 577 | 77 | ||
Set Membership: |
|