Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01202064_g1
          See other TRIM56 GE Assays ›
          Gene Symbol:
          Trim56
          Gene Name:
          tripartite motif-containing 56
          Gene Aliases:
          A130009K11Rik, Gm452, RNF109
          Chromosome Location:
          Chr.5: 137111286 - 137116466 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01202064_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trim56
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_201373.4 NP_958761.1 2 - 3 261 72
          GenBank mRNA AK037350.1 - 2 - 3 262 72
          BC045615.1 - 2 - 3 251 4976515 72

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK037350.1, BC045615.1, NM_201373.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trim56

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          interferon-beta production
          response to type I interferon
          innate immune response
          negative regulation of viral entry into host cell
          defense response to virus
          protein K63-linked ubiquitination
          negative regulation of viral release from host cell
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          poly(A) RNA binding
          metal ion binding
          ubiquitin protein ligase activity
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139763 BC045615.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724708 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346584 BC039782.1 genbank_mrna_mouse
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106283 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139763 BC045615.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P244820 BC045615.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P244820 AK037350.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P244820 NM_201373.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124676 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05759854 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106283 - -
          Whole Mouse Genome G4122A A_51_P244820 BC045615.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P244820 AK037350.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P244820 NM_201373.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129681 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112484 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106283 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04321859 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04593665 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04593668 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04593667 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04321860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04668643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04668640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04575903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04399426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04679998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04672990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04668641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04321804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04497969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04672975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04678755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04679995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04672973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04679994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04123614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04904734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04392916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04679997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124567 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534682 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534681 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534680 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534683 - -
          Affymetrix Mouse Gene 1.1 ST Array 10534679 - -
          Affymetrix Mouse Clariom S Array PSR0500031113.mm.1 NM_201373.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031113.mm.1 NM_201373.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031113.mm.1 TC0500003433.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031115.mm.1 NM_201373.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031115.mm.1 NM_201373.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031115.mm.1 TC0500003433.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031116.mm.1 NM_201373.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031116.mm.1 NM_201373.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031116.mm.1 TC0500003433.mm.1 MTA TC
          Murine Genome U74v2 Set 131166_PM_at - -
          Murine Genome U74v2 Set 131166_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 687527 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1580528 - -
          Illumina mouse-6 v1.0 expression beadchip 1580528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001580528 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2441921 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2441921 - -
          mouse-6 v1.1 (Illumina) scl0384309.1_171-S - -
          Illumina Sentrix mouse-6.v1 bead array 1580528 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0384309.1_171-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580528 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2441921 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2441921 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline