Search
Search
| Gene Symbol: Cdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271566.1 | NP_001258495.1 | 2 - 3 | 457 | 141 | |
| NM_001271567.1 | NP_001258496.1 | 2 - 3 | 288 | 141 | ||
| NM_001271568.1 | NP_001258497.1 | 2 - 3 | 288 | 141 | ||
| NM_009863.3 | NP_033993.2 | 2 - 3 | 288 | 141 | ||
| GenBank mRNA | AB018576.1 | - | 1 - 2 | 99 | 141 | |
| AB018577.1 | - | 1 - 2 | 99 | 141 | ||
| AJ007661.1 | - | 2 - 3 | 240 | 141 | ||
| AK012986.1 | - | 2 - 3 | 271 | 141 | ||
| AK145921.1 | - | 2 - 3 | 214 | 141 | ||
| AK146009.1 | - | 2 - 3 | 459 | 141 | ||
| AK163921.1 | - | 258 | 141 | |||
| BC062149.1 | - | 2 - 3 | 218 | 141 | ||
| BC072666.1 | - | 2 - 3 | 229 | 30611704 | 141 | |
| BC080702.1 | - | 2 - 3 | 241 | 30474846 | 141 | |
Set Membership: |
|