Search
Search

Usp42| Gene Symbol: Usp42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029749.2 | NP_084025.2 | 5 - 6 | 778 | 61 | |
| GenBank mRNA | AK010801.1 | - | 4 - 5 | 588 | 61 | |
| AY336567.1 | - | 4 - 5 | 651 | 61 | ||
| BC137852.1 | - | 4 - 5 | 671 | 8860871 | 61 | |
| BG079818.2 | - | 1 - 2 | 60 | 61 | ||
Set Membership: |
|