Search
Search
| Gene Symbol: Oas3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145226.2 | NP_660261.1 | 6 - 7 | 1568 | 127 | |
| GenBank mRNA | AB067534.1 | - | 6 - 7 | 1568 | 127 | |
| AF453830.1 | - | 6 - 7 | 1560 | 127 | ||
| AK158392.1 | - | 6 - 7 | 1551 | 127 | ||
| BC141055.1 | - | 6 - 7 | 1561 | 9055667 | 127 | |
Set Membership: |
|