Search
Search

Cdc37l1| Gene Symbol: Cdc37l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012312.1 | - | 2 - 3 | 451 | 151 | |
| AK016651.1 | - | 2 - 3 | 594 | 151 | ||
| AK038578.1 | - | 2 - 3 | 415 | 152 | ||
| AK044291.1 | - | 2 - 3 | 564 | 151 | ||
| AK048044.1 | - | 2 - 3 | 564 | 151 | ||
| AK076231.1 | - | 2 - 3 | 578 | 151 | ||
| AK078139.1 | - | 2 - 3 | 562 | 151 | ||
| AK078468.1 | - | 2 - 3 | 578 | 151 | ||
| AK079556.1 | - | 2 - 3 | 415 | 151 | ||
| AK080238.1 | - | 2 - 3 | 460 | 151 | ||
| AK161465.1 | - | 2 - 3 | 473 | 151 | ||
| AK165398.1 | - | 2 - 3 | 580 | 151 | ||
| BC031761.1 | - | 2 - 3 | 444 | 4235549 | 151 | |
Set Membership: |
|