Search
Search

Flt3l| Gene Symbol: Flt3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013520.3 | NP_038548.3 | 4 - 5 | 293 | 64 | |
| NR_102725.1 | 4 - 5 | 293 | 64 | |||
| GenBank mRNA | AK020105.1 | - | 4 - 5 | 293 | 64 | |
| BC019801.1 | - | 4 - 5 | 278 | 5132319 | 64 | |
| CK622267.1 | - | 4 - 5 | 263 | 64 | ||
| EU274583.1 | - | 3 - 4 | 205 | 64 | ||
| L23636.1 | - | 3 - 4 | 236 | 64 | ||
| S76459.1 | - | 3 - 4 | 211 | 64 | ||
| U04807.1 | - | 3 - 4 | 460 | 64 | ||
Set Membership: |
|